The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006943.1 | NP_001006944.1 | 2 - 3 | 287 | 59 |
XM_011540970.1 | XP_011539272.1 | 2 - 3 | 184 | 59 | |
GenBank mRNA | BC038796.1 | - | 2 - 3 | 217 | 59 |
BC072417.1 | - | 2 - 3 | 287 | 59 | |
DR007053.1 | - | 2 - 3 | 297 | 59 |