Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPHA8
Entrez Gene ID
2046
Gene Name
EPH receptor A8
Gene Aliases
EEK, EK3, HEK3
Chromosome Location
Chr.1: 22563508 - 22603594 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011540969.1 XP_011539271.1 10 - 11 1983 59
XM_011540970.1 XP_011539272.1 8 - 9 1717 59
XM_011540972.1 XP_011539274.1 9 - 10 1884 59
XM_011540973.1 XP_011539275.1 7 - 8 1619 59
XM_011540975.2 XP_011539277.1 5 - 6 795 59
GenBank mRNA AB040892.1 - 6 - 7 1241 59

View Full Product Details