The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010898.2 | NP_001010898.1 | 3 - 4 | 905 | 108 |
XM_006710643.2 | XP_006710706.1 | 3 - 4 | 613 | 108 | |
GenBank mRNA | AK289982.1 | - | 3 - 4 | 901 | 108 |
BC140908.1 | - | 2 - 3 | 490 | 108 |