The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170629.1 | NP_001164100.1 | 23 - 24 | 4794 | 99 |
NM_020920.3 | NP_065971.2 | 24 - 25 | 4039 | 99 | |
GenBank mRNA | AB046784.3 | - | 23 - 24 | 4282 | 99 |
AK056639.1 | - | 4 - 5 | 661 | 99 | |
AK122910.1 | - | 24 - 25 | 3987 | 99 | |
AK131077.1 | - | 9 - 10 | 1618 | 99 | |
CR749315.1 | - | 24 - 25 | 3984 | 99 |