The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170629.1 | NP_001164100.1 | 28 - 29 | 5243 | 82 |
NM_020920.3 | NP_065971.2 | 29 - 30 | 4488 | 82 | |
GenBank mRNA | AB046784.3 | - | 28 - 29 | 4731 | 82 |
AK056639.1 | - | 9 - 10 | 1110 | 82 | |
AK122910.1 | - | 29 - 30 | 4436 | 82 | |
AK131077.1 | - | 14 - 15 | 2067 | 82 | |
BC098452.1 | - | 3 - 4 | 159 | 82 | |
CR749315.1 | - | 29 - 30 | 4433 | 82 | |
U00930.1 | - | - | 82 |