The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170629.1 | NP_001164100.1 | 3 - 4 | 1663 | 107 |
NM_020920.3 | NP_065971.2 | 4 - 5 | 908 | 107 | |
GenBank mRNA | AB046784.3 | - | 3 - 4 | 1151 | 107 |
CR749315.1 | - | 4 - 5 | 853 | 107 |