Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDR2
Entrez Gene ID
4921
Gene Name
discoidin domain receptor tyrosine kinase 2
Gene Aliases
MIG20a, NTRKR3, TKT, TYRO10
Chromosome Location
Chr.1: 162631213 - 162783524 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014796.1 NP_001014796.1 10 - 11 1569 63
NM_006182.2 NP_006173.2 9 - 10 1489 63
XM_006711344.3 XP_006711407.1 9 - 10 1530 63
XM_011509586.2 XP_011507888.1 9 - 10 1462 63
XM_011509587.2 XP_011507889.1 10 - 11 1523 63
XM_011509588.2 XP_011507890.1 9 - 10 1526 63
GenBank mRNA AK225063.1 - 9 - 10 1408 63
AK314388.1 - 9 - 10 1438 63
BC052998.2 - 10 - 11 1540 63
BX640899.1 - 10 - 11 1526 63
X74764.1 - 9 - 10 1457 63

View Full Product Details