The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153240.4 | NP_694972.3 | 13 - 14 | 2088 | 110 |
GenBank mRNA | AK055253.1 | - | 8 - 9 | 1031 | 110 |
AK092910.1 | - | 3 - 4 | 367 | 110 | |
AL832863.1 | - | 3 - 4 | 322 | 110 | |
AY257864.1 | - | 13 - 14 | 2017 | 110 |