Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCSK1
Entrez Gene ID
5122
Gene Name
proprotein convertase subtilisin/kexin type 1
Gene Aliases
BMIQ12, NEC1, PC1, PC3, SPC3
Chromosome Location
Chr.5: 96390336 - 96433281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000439.4 NP_000430.3 6 - 7 944 105
NM_001177875.1 NP_001171346.1 6 - 7 602 105
GenBank mRNA AB208874.1 - 6 - 7 620 105
AK303888.1 - 6 - 7 602 105
BC130295.1 - 6 - 7 826 105
BC136486.1 - 6 - 7 826 105
M90753.1 - 6 - 7 914 105
X64810.1 - 6 - 7 894 105

View Full Product Details