The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000730.2 | NP_000721.1 | 1 - 2 | 267 | 90 |
GenBank mRNA | AK313978.1 | - | 1 - 2 | 309 | 90 |
AY322549.1 | - | 1 - 2 | 114 | 90 | |
BC074987.2 | - | 1 - 2 | 162 | 90 | |
L13605.1 | - | 1 - 2 | 185 | 90 | |
L19315.1 | - | 1 - 2 | 267 | 90 |