The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000160.4 | NP_000151.1 | 9 - 10 | 1170 | 83 |
XM_006722277.1 | XP_006722340.1 | 9 - 10 | 1199 | 83 | |
XM_011523539.1 | XP_011521841.1 | 9 - 10 | 1297 | 83 | |
XM_017024446.1 | XP_016879935.1 | 9 - 10 | 1218 | 83 | |
XM_017024447.1 | XP_016879936.1 | 8 - 9 | 1007 | 83 | |
GenBank mRNA | AK314247.1 | - | 8 - 9 | 1243 | 83 |
BC104854.1 | - | 8 - 9 | 920 | 83 | |
BC112041.1 | - | 8 - 9 | 919 | 83 | |
L20316.1 | - | 9 - 10 | 1152 | 83 | |
U03469.1 | - | 8 - 9 | 936 | 83 |