Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GC
Entrez Gene ID
2638
Gene Name
GC, vitamin D binding protein
Gene Aliases
DBP, DBP/GC, GRD3, Gc-MAF, GcMAF, HEL-S-51, VDBG, VDBP
Chromosome Location
Chr.4: 71741693 - 71805520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000583.3 NP_000574.2 7 - 8 1177 149
NM_001204306.1 NP_001191235.1 8 - 9 1048 149
NM_001204307.1 NP_001191236.1 8 - 9 985 149
XM_006714177.2 XP_006714240.1 7 - 8 1040 149
GenBank mRNA AK223458.1 - 8 - 9 1048 149
AK290827.1 - 7 - 8 887 149
AK298433.1 - 6 - 7 745 149
AK309595.1 - 8 - 9 985 149
AK312971.1 - 7 - 8 891 149
AK315853.1 - 7 - 8 1040 149
BC057228.1 - 7 - 8 895 149
EU794674.1 - 7 - 8 986 149
X03178.1 - 7 - 8 986 149

View Full Product Details