The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020137.4 | NP_064522.3 | 15 - 16 | 1312 | 83 |
XM_017029656.1 | XP_016885145.1 | 15 - 16 | 1312 | 83 | |
XM_017029658.1 | XP_016885147.1 | 15 - 16 | 2622 | 83 | |
XM_017029659.1 | XP_016885148.1 | 15 - 16 | 1359 | 83 | |
XM_017029660.1 | XP_016885149.1 | 15 - 16 | 1713 | 83 | |
XM_017029661.1 | XP_016885150.1 | 15 - 16 | 1359 | 83 | |
GenBank mRNA | AB032993.1 | - | 15 - 16 | 1257 | 83 |
AK292158.1 | - | 15 - 16 | 1283 | 83 | |
AL136847.1 | - | 2 - 3 | 198 | 83 | |
BC001522.2 | - | 5 - 6 | 425 | 83 | |
BC101544.1 | - | 15 - 16 | 1310 | 83 | |
BC101546.1 | - | 15 - 16 | 1310 | 83 | |
BX640704.1 | - | 9 - 10 | 996 | 83 | |
BX647801.1 | - | 14 - 15 | 1152 | 83 | |
BX647804.1 | - | 14 - 15 | 1152 | 83 |