The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020137.4 | NP_064522.3 | 23 - 24 | 2215 | 70 |
XM_011543935.1 | XP_011542237.1 | 23 - 24 | 2245 | 70 | |
XM_011543936.1 | XP_011542238.1 | 22 - 23 | 2167 | 70 | |
XM_011543937.1 | XP_011542239.1 | 23 - 24 | 2292 | 70 | |
XM_017029656.1 | XP_016885145.1 | 22 - 23 | 2137 | 70 | |
XM_017029657.1 | XP_016885146.1 | 23 - 24 | 2616 | 70 | |
XM_017029658.1 | XP_016885147.1 | 23 - 24 | 3525 | 70 | |
XM_017029659.1 | XP_016885148.1 | 23 - 24 | 2262 | 70 | |
XM_017029660.1 | XP_016885149.1 | 22 - 23 | 2538 | 70 | |
XM_017029661.1 | XP_016885150.1 | 22 - 23 | 2184 | 70 | |
GenBank mRNA | AB032993.1 | - | 23 - 24 | 2160 | 70 |
AJ297364.1 | - | 2 - 3 | 152 | 70 | |
AK292158.1 | - | 23 - 24 | 2186 | 70 | |
AL136847.1 | - | 10 - 11 | 1101 | 70 | |
BC001522.2 | - | 13 - 14 | 1328 | 70 | |
BC101544.1 | - | 23 - 24 | 2213 | 70 | |
BC101546.1 | - | 23 - 24 | 2213 | 70 |