Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPAS1
Entrez Gene ID
4861
Gene Name
neuronal PAS domain protein 1
Gene Aliases
MOP5, PASD5, bHLHe11
Chromosome Location
Chr.19: 47019820 - 47045775 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002517.3 NP_002508.2 3 - 4 549 149
XM_006723231.2 XP_006723294.1 3 - 4 573 149
XM_011526997.1 XP_011525299.1 3 - 4 573 149
XM_011526999.1 XP_011525301.1 3 - 4 573 149
XM_011527001.1 XP_011525303.1 3 - 4 573 149
XM_011527002.1 XP_011525304.1 3 - 4 573 149
XM_011527006.1 XP_011525308.1 3 - 4 573 149
XM_011527007.1 XP_011525309.1 3 - 4 573 149
XM_017026841.1 XP_016882330.1 3 - 4 573 149
XM_017026842.1 XP_016882331.1 3 - 4 573 149
XM_017026843.1 XP_016882332.1 3 - 4 573 149
GenBank mRNA AB054002.1 - 2 - 3 514 149
BC039016.1 - 3 - 4 549 149
U77968.1 - 2 - 3 377 149

View Full Product Details