Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPATCH1
Entrez Gene ID
55094
Gene Name
G-patch domain containing 1
Gene Aliases
ECGP, GPATC1
Chromosome Location
Chr.19: 33080880 - 33130412 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018025.2 NP_060495.2 10 - 11 1593 109
NR_135270.1 10 - 11 1593 109
XM_006723255.3 XP_006723318.1 10 - 11 1308 109
GenBank mRNA AF434677.1 - 10 - 11 1279 109
AK096005.1 - 10 - 11 1287 109
AK292503.1 - 10 - 11 1287 109
AK315712.1 - 10 - 11 1305 109
AL834465.1 - 10 - 11 1284 109
BC006108.1 - 10 - 11 1593 109

View Full Product Details