The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018025.2 | NP_060495.2 | 14 - 15 | 2341 | 152 |
NR_135270.1 | 14 - 15 | 2341 | 152 | ||
GenBank mRNA | AF434677.1 | - | 14 - 15 | 2027 | 152 |
AK001068.1 | - | 4 - 5 | 728 | 152 | |
AK091835.1 | - | 1 - 2 | 963 | 152 | |
AK096005.1 | - | 14 - 15 | 2035 | 152 | |
AK292503.1 | - | 14 - 15 | 2035 | 152 | |
AK315712.1 | - | 14 - 15 | 2053 | 152 | |
AL834465.1 | - | 14 - 15 | 2032 | 152 | |
BC006108.1 | - | 14 - 15 | 2341 | 152 |