Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPATCH1
Entrez Gene ID
55094
Gene Name
G-patch domain containing 1
Gene Aliases
ECGP, GPATC1
Chromosome Location
Chr.19: 33080880 - 33130412 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018025.2 NP_060495.2 3 - 4 608 90
NR_135270.1 3 - 4 608 90
XM_006723255.3 XP_006723318.1 3 - 4 323 90
GenBank mRNA AF434677.1 - 3 - 4 294 90
AK096005.1 - 3 - 4 302 90
AK292503.1 - 3 - 4 302 90
AK315712.1 - 3 - 4 320 90
AL834465.1 - 3 - 4 299 90
BC006108.1 - 3 - 4 608 90

View Full Product Details