Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL4
Entrez Gene ID
56062
Gene Name
kelch like family member 4
Gene Aliases
DKELCHL, KHL4
Chromosome Location
Chr.X: 87517541 - 87670050 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019117.4 NP_061990.2 1 - 2 597 78
NM_057162.2 NP_476503.1 1 - 2 597 78
GenBank mRNA AB051474.1 - 1 - 2 543 78
AF284765.1 - 1 - 2 504 78
AF284766.1 - 1 - 2 504 78
AK056457.1 - 1 - 2 565 78
AL832514.1 - 1 - 2 537 78
BC140839.1 - 1 - 2 466 78
BC140840.1 - 1 - 2 466 78
BC142652.1 - 1 - 2 443 78
BC146677.1 - 1 - 2 443 78
BC161675.1 - 1 - 2 543 78

View Full Product Details