Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TYW5
Entrez Gene ID
129450
Gene Name
tRNA-yW synthesizing protein 5
Gene Aliases
C2orf60, hTYW5
Chromosome Location
Chr.2: 199928911 - 199955736 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039693.2 NP_001034782.1 3 - 4 576 77
NR_004862.1 3 - 4 557 77
NR_109906.1 3 - 4 557 77
NR_109907.1 2 - 3 421 77
GenBank mRNA AK095272.1 - 2 - 3 185 77
AK293064.1 - 3 - 4 324 77
AK298977.1 - 3 - 4 320 77
BC025758.1 - 3 - 4 304 77
BC132835.1 - 3 - 4 340 77
BC136837.1 - 3 - 4 340 77
BX482572.1 - 3 - 4 330 77
BX647677.1 - 3 - 4 558 77
CN261486.1 - 3 - 4 289 77

View Full Product Details