The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040428.3 | NP_001035518.1 | 10 - 11 | 1286 | 89 |
NM_018418.4 | NP_060888.2 | 11 - 12 | 1382 | 89 | |
XM_005267851.1 | XP_005267908.1 | 11 - 12 | 1356 | 89 | |
XM_005267852.1 | XP_005267909.1 | 10 - 11 | 1285 | 89 | |
XM_005267854.1 | XP_005267911.1 | 12 - 13 | 1418 | 89 | |
XM_005267855.1 | XP_005267912.1 | 11 - 12 | 1351 | 89 | |
XM_006720204.1 | XP_006720267.1 | 11 - 12 | 1356 | 89 | |
XM_006720205.1 | XP_006720268.1 | 11 - 12 | 1356 | 89 | |
XM_011536951.1 | XP_011535253.1 | 11 - 12 | 1550 | 89 | |
XM_011536952.1 | XP_011535254.1 | 9 - 10 | 1168 | 89 | |
XM_011536953.1 | XP_011535255.1 | 10 - 11 | 1489 | 89 | |
XM_017021452.1 | XP_016876941.1 | 11 - 12 | 1547 | 89 | |
XM_017021453.1 | XP_016876942.1 | 11 - 12 | 1516 | 89 | |
XM_017021454.1 | XP_016876943.1 | 12 - 13 | 1415 | 89 | |
XM_017021455.1 | XP_016876944.1 | 11 - 12 | 1348 | 89 | |
XM_017021456.1 | XP_016876945.1 | 12 - 13 | 1361 | 89 | |
XM_017021457.1 | XP_016876946.1 | 10 - 11 | 1486 | 89 | |
GenBank mRNA | AF144487.3 | - | 11 - 12 | 1362 | 89 |
AF144488.2 | - | 9 - 10 | 1090 | 89 | |
AK055864.1 | - | 10 - 11 | 2954 | 89 | |
AK225746.1 | - | 12 - 13 | 1356 | 89 | |
AK290631.1 | - | 10 - 11 | 1257 | 89 | |
BC008656.1 | - | 6 - 7 | 814 | 89 | |
BC090875.1 | - | 11 - 12 | 1669 | 89 | |
CR936777.1 | - | 11 - 12 | 1328 | 89 | |
EU668353.1 | - | 11 - 12 | 1358 | 89 |