Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SPATA7
- Entrez Gene ID
-
55812
- Gene Name
- spermatogenesis associated 7
- Gene Aliases
-
HEL-S-296, HSD-3.1, HSD3, LCA3
- Chromosome Location
-
Chr.14: 88385638 - 88470350 on Build GRCh38
View Full Product Details