The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039877.1 | NP_001034966.1 | 14 - 15 | 1960 | 98 |
NM_013403.2 | NP_037535.2 | 14 - 15 | 1939 | 98 | |
XM_006723171.2 | XP_006723234.1 | 15 - 16 | 2017 | 98 | |
XM_006723172.2 | XP_006723235.1 | 15 - 16 | 1996 | 98 | |
XM_006723174.2 | XP_006723237.1 | 14 - 15 | 1767 | 98 | |
XM_011526878.1 | XP_011525180.1 | 15 - 16 | 2141 | 98 | |
XM_011526879.1 | XP_011525181.1 | 8 - 9 | 1104 | 98 | |
XM_017026716.1 | XP_016882205.1 | 15 - 16 | 1746 | 98 | |
XM_017026717.1 | XP_016882206.1 | 14 - 15 | 1722 | 98 | |
XM_017026718.1 | XP_016882207.1 | 10 - 11 | 1511 | 98 | |
XM_017026719.1 | XP_016882208.1 | 14 - 15 | 2096 | 98 | |
GenBank mRNA | AB209003.1 | - | 14 - 15 | 1912 | 98 |
AF212940.1 | - | 14 - 15 | 1906 | 98 | |
AK056294.1 | - | 8 - 9 | 961 | 98 | |
AK092913.1 | - | 5 - 6 | 1275 | 98 | |
AK225493.1 | - | 8 - 9 | 935 | 98 | |
AK297637.1 | - | 15 - 16 | 1918 | 98 | |
AK308813.1 | - | 9 - 10 | 1174 | 98 | |
BC004910.2 | - | 6 - 7 | 694 | 98 | |
BC034604.1 | - | 14 - 15 | 1916 | 98 |