Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBRS
Entrez Gene ID
64319
Gene Name
fibrosin
Gene Aliases
FBS, FBS1
Chromosome Location
Chr.16: 30658142 - 30670810 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105079.2 NP_001098549.2 16 - 17 2155 103
XM_011545916.2 XP_011544218.1 15 - 16 3502 103
XM_011545917.2 XP_011544219.1 15 - 16 3502 103
XM_011545919.2 XP_011544221.1 14 - 15 3465 103
GenBank mRNA AK021680.1 - 4 - 5 346 103
AK022551.1 - 7 - 8 560 103
AK096322.1 - 8 - 9 938 103
AK298229.1 - 9 - 10 718 103
BQ276414.1 - 9 - 10 723 103
DR761499.1 - 9 - 10 652 103

View Full Product Details