Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBRS
Entrez Gene ID
64319
Gene Name
fibrosin
Gene Aliases
FBS, FBS1
Chromosome Location
Chr.16: 30658142 - 30670810 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105079.2 NP_001098549.2 17 - 18 2366 83
XM_011545916.2 XP_011544218.1 16 - 17 3713 83
XM_011545917.2 XP_011544219.1 16 - 17 3713 83
XM_011545919.2 XP_011544221.1 15 - 16 3676 83
GenBank mRNA AK021680.1 - 5 - 6 557 83
AK022551.1 - 8 - 9 771 83
AK096322.1 - 9 - 10 1149 83

View Full Product Details