The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177983.2 | NP_817092.1 | 2 - 3 | 448 | 68 |
GenBank mRNA | AI879284.1 | - | 2 - 3 | 398 | 68 |
AK127593.1 | - | 2 - 3 | 314 | 68 | |
AK293141.1 | - | 3 - 4 | 242 | 68 | |
BC000057.2 | - | 2 - 3 | 297 | 68 | |
BC022061.2 | - | 2 - 3 | 294 | 68 | |
BT007454.1 | - | 2 - 3 | 187 | 68 | |
CR457059.1 | - | 2 - 3 | 187 | 68 | |
DA520893.1 | - | 2 - 3 | 448 | 68 | |
Y13936.1 | - | 2 - 3 | 211 | 68 |