The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177983.2 | NP_817092.1 | 8 - 9 | 1589 | 79 |
GenBank mRNA | AB209197.1 | - | 6 - 7 | 3035 | 79 |
AK127593.1 | - | 8 - 9 | 1455 | 79 | |
AK293141.1 | - | 9 - 10 | 1383 | 79 | |
AK312455.1 | - | 8 - 9 | 1579 | 79 | |
BC000057.2 | - | 8 - 9 | 1438 | 79 | |
BC007361.2 | - | 4 - 5 | 690 | 79 | |
BC022061.2 | - | 8 - 9 | 1435 | 79 | |
BT007454.1 | - | 8 - 9 | 1328 | 79 | |
CR457059.1 | - | 8 - 9 | 1328 | 79 | |
Y13936.1 | - | 8 - 9 | 1352 | 79 |