Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIF5C
Entrez Gene ID
3800
Gene Name
kinesin family member 5C
Gene Aliases
CDCBM2, KINN, NKHC, NKHC-2, NKHC2
Chromosome Location
Chr.2: 148875223 - 149026759 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004522.2 NP_004513.1 20 - 21 2706 69
NR_111932.1 13 - 14 1728 69
XM_011511157.2 XP_011509459.1 13 - 14 1787 69
XM_017004062.1 XP_016859551.1 20 - 21 2697 69
GenBank mRNA AB011103.1 - 20 - 21 2679 69
AB209191.1 - 1 - 2 589 69
BC110287.1 - 13 - 14 1658 69
BC127772.1 - 17 - 18 2030 69

View Full Product Details