The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004522.2 | NP_004513.1 | 21 - 22 | 2775 | 71 |
NR_111932.1 | 14 - 15 | 1797 | 71 | ||
XM_011511157.2 | XP_011509459.1 | 14 - 15 | 1856 | 71 | |
XM_017004062.1 | XP_016859551.1 | 21 - 22 | 2766 | 71 | |
GenBank mRNA | AB011103.1 | - | 21 - 22 | 2748 | 71 |
AB209191.1 | - | 2 - 3 | 658 | 71 | |
BC110287.1 | - | 14 - 15 | 1727 | 71 | |
BC127772.1 | - | 18 - 19 | 2099 | 71 |