The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004522.2 | NP_004513.1 | 22 - 23 | 2843 | 110 |
NR_111932.1 | 15 - 16 | 1865 | 110 | ||
XM_011511157.2 | XP_011509459.1 | 15 - 16 | 1924 | 110 | |
XM_017004062.1 | XP_016859551.1 | 22 - 23 | 2834 | 110 | |
GenBank mRNA | AB011103.1 | - | 22 - 23 | 2816 | 110 |
AB209191.1 | - | 3 - 4 | 726 | 110 | |
BC110287.1 | - | 15 - 16 | 1795 | 110 | |
BC127772.1 | - | 19 - 20 | 2167 | 110 |