The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004522.2 | NP_004513.1 | 23 - 24 | 2945 | 74 |
NR_111932.1 | 16 - 17 | 1967 | 74 | ||
XM_011511157.2 | XP_011509459.1 | 16 - 17 | 2026 | 74 | |
XM_017004062.1 | XP_016859551.1 | 23 - 24 | 2936 | 74 | |
GenBank mRNA | AB011103.1 | - | 23 - 24 | 2918 | 74 |
AB209191.1 | - | 4 - 5 | 828 | 74 | |
BC110287.1 | - | 16 - 17 | 1897 | 74 | |
BC127772.1 | - | 20 - 21 | 2269 | 74 |