The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016445.2 | NP_057529.1 | 2 - 3 | 347 | 82 |
XM_011536644.2 | XP_011534946.1 | 2 - 3 | 321 | 82 | |
GenBank mRNA | AF228603.1 | - | 2 - 3 | 258 | 82 |
BC001226.1 | - | 2 - 3 | 291 | 82 | |
BC008056.2 | - | 2 - 3 | 201 | 82 | |
CR976852.1 | - | 2 - 3 | 351 | 82 |