Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEK2
Entrez Gene ID
26499
Gene Name
pleckstrin 2
Gene Aliases
-
Chromosome Location
Chr.14: 67386979 - 67412200 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016445.2 NP_057529.1 6 - 7 916 66
GenBank mRNA AF228603.1 - 6 - 7 827 66
BC001226.1 - 6 - 7 860 66
BC008056.2 - 6 - 7 770 66
BX114063.1 - 1 - 2 46 66

View Full Product Details