The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015529.3 | NP_056344.2 | 6 - 7 | 1046 | 121 |
XM_017010714.1 | XP_016866203.1 | 6 - 7 | 854 | 121 | |
XM_017010715.1 | XP_016866204.1 | 6 - 7 | 692 | 121 | |
GenBank mRNA | AK074879.1 | - | 6 - 7 | 761 | 121 |
AK226054.1 | - | 2 - 3 | 143 | 121 | |
AL080058.1 | - | 1 - 2 | 104 | 121 | |
AY007239.1 | - | 5 - 6 | 962 | 121 | |
AY359094.1 | - | 6 - 7 | 970 | 121 | |
BC018756.1 | - | 6 - 7 | 1005 | 121 |