The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001901.2 | NP_001892.1 | 2 - 3 | 501 | 66 |
GenBank mRNA | AK092280.1 | - | 2 - 3 | 352 | 66 |
AK125220.1 | - | 2 - 3 | 286 | 66 | |
AK290884.1 | - | 2 - 3 | 495 | 66 | |
AK315207.1 | - | 2 - 3 | 495 | 66 | |
AY395801.1 | - | 2 - 3 | 418 | 66 | |
AY550024.1 | - | 2 - 3 | 408 | 66 | |
BC087839.1 | - | 2 - 3 | 477 | 66 | |
BT019794.1 | - | 2 - 3 | 295 | 66 | |
BT019795.1 | - | 2 - 3 | 295 | 66 | |
CR541734.1 | - | 2 - 3 | 295 | 66 | |
CR541759.1 | - | 2 - 3 | 295 | 66 | |
M92934.1 | - | 2 - 3 | 424 | 66 | |
U14750.1 | - | 1 - 2 | 75 | 66 | |
X78947.1 | - | 2 - 3 | 440 | 66 |