The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079521.1 | NP_001072989.1 | 20 - 21 | 2469 | 129 |
NM_001079522.1 | NP_001072990.1 | 21 - 22 | 2593 | 129 | |
NM_001271014.1 | NP_001257943.1 | 21 - 22 | 2593 | 129 | |
NM_004986.3 | NP_004977.2 | 20 - 21 | 2469 | 129 | |
NR_073128.1 | 21 - 22 | 2593 | 129 | ||
NR_073129.1 | 21 - 22 | 2593 | 129 | ||
XM_006720138.2 | XP_006720201.1 | 21 - 22 | 2584 | 129 | |
XM_006720139.2 | XP_006720202.1 | 20 - 21 | 2432 | 129 | |
XM_006720140.3 | XP_006720203.1 | 20 - 21 | 2432 | 129 | |
XM_006720141.2 | XP_006720204.1 | 20 - 21 | 2432 | 129 | |
XM_011536751.2 | XP_011535053.1 | 20 - 21 | 2690 | 129 | |
XM_011536752.1 | XP_011535054.1 | 20 - 21 | 2432 | 129 | |
XM_011536753.1 | XP_011535055.1 | 20 - 21 | 2432 | 129 | |
XM_017021278.1 | XP_016876767.1 | 20 - 21 | 8282 | 129 | |
XM_017021279.1 | XP_016876768.1 | 20 - 21 | 2432 | 129 | |
XM_017021280.1 | XP_016876769.1 | 21 - 22 | 2584 | 129 | |
XM_017021281.1 | XP_016876770.1 | 21 - 22 | 8406 | 129 | |
XM_017021282.1 | XP_016876771.1 | 21 - 22 | 2587 | 129 | |
XM_017021283.1 | XP_016876772.1 | 21 - 22 | 2521 | 129 | |
XM_017021284.1 | XP_016876773.1 | 20 - 21 | 2432 | 129 | |
XM_017021285.1 | XP_016876774.1 | 21 - 22 | 2564 | 129 | |
XM_017021286.1 | XP_016876775.1 | 20 - 21 | 2432 | 129 | |
GenBank mRNA | AK302797.1 | - | 21 - 22 | 2547 | 129 |
AK308272.1 | - | 3 - 4 | 288 | 129 | |
AY264265.1 | - | 21 - 22 | 2478 | 129 | |
AY536375.1 | - | 10 - 11 | 682 | 129 | |
BC050555.1 | - | 20 - 21 | 2408 | 129 | |
BC112337.1 | - | 21 - 22 | 2476 | 129 | |
BC117132.1 | - | 20 - 21 | 2410 | 129 | |
BC143720.1 | - | 20 - 21 | 2410 | 129 | |
D13629.1 | - | 20 - 21 | 2374 | 129 | |
L25616.1 | - | 19 - 20 | 2331 | 129 | |
Z22551.1 | - | 20 - 21 | 2357 | 129 |