The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142545.1 | NP_001136017.1 | 1 - 2 | 90 | 111 |
NM_020421.3 | NP_065154.2 | 1 - 2 | 90 | 111 | |
XM_005267885.3 | XP_005267942.1 | 1 - 2 | 113 | 111 | |
XM_005267886.3 | XP_005267943.1 | 1 - 2 | 109 | 111 | |
XM_011536987.2 | XP_011535289.1 | 1 - 2 | 109 | 111 | |
XM_011536988.2 | XP_011535290.1 | 1 - 2 | 109 | 111 | |
XM_017021474.1 | XP_016876963.1 | 1 - 2 | 109 | 111 | |
XM_017021475.1 | XP_016876964.1 | 1 - 2 | 109 | 111 | |
GenBank mRNA | CD630155.1 | - | 1 - 2 | 95 | 111 |