Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADCK1
Entrez Gene ID
57143
Gene Name
aarF domain containing kinase 1
Gene Aliases
-
Chromosome Location
Chr.14: 77800058 - 77933954 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142545.1 NP_001136017.1 2 - 3 235 117
NM_020421.3 NP_065154.2 2 - 3 235 117
XM_005267885.3 XP_005267942.1 2 - 3 258 117
XM_005267886.3 XP_005267943.1 2 - 3 254 117
XM_011536987.2 XP_011535289.1 2 - 3 254 117
XM_011536988.2 XP_011535290.1 2 - 3 254 117
XM_017021474.1 XP_016876963.1 2 - 3 254 117
XM_017021475.1 XP_016876964.1 2 - 3 254 117
XM_017021476.1 XP_016876965.1 2 - 3 182 117
GenBank mRNA AK096919.1 - 2 - 3 181 117
AK299430.1 - 2 - 3 198 117
BC017459.1 - 2 - 3 192 117
BC058906.1 - 2 - 3 187 117
BX248292.1 - 2 - 3 197 117
CD630155.1 - 2 - 3 240 117
DN996748.1 - 2 - 3 189 117

View Full Product Details