Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAPN15
Entrez Gene ID
6650
Gene Name
calpain 15
Gene Aliases
SOLH
Chromosome Location
Chr.16: 527795 - 554636 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005632.2 NP_005623.1 10 - 11 2869 85
XM_011522620.2 XP_011520922.1 11 - 12 6168 85
XM_011522621.2 XP_011520923.1 11 - 12 7096 85
XM_011522622.1 XP_011520924.1 9 - 10 5800 85
XM_011522623.2 XP_011520925.1 12 - 13 3871 85
XM_011522624.2 XP_011520926.1 11 - 12 3209 85
XM_011522625.2 XP_011520927.1 10 - 11 3095 85
XM_011522626.2 XP_011520928.1 11 - 12 6660 85
XM_011522627.2 XP_011520929.1 11 - 12 6665 85
XM_011522628.2 XP_011520930.1 11 - 12 3729 85
XM_011522629.2 XP_011520931.1 11 - 12 3592 85
XM_011522630.2 XP_011520932.1 10 - 11 2930 85
XM_011522631.2 XP_011520933.1 9 - 10 2816 85
XM_011522632.2 XP_011520934.1 9 - 10 3247 85
XM_017023596.1 XP_016879085.1 12 - 13 4008 85
GenBank mRNA BC021854.1 - 6 - 7 936 85
U85647.1 - 10 - 11 2869 85

View Full Product Details