The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005632.2 | NP_005623.1 | 5 - 6 | 2023 | 148 |
XM_011522620.2 | XP_011520922.1 | 6 - 7 | 5322 | 148 | |
XM_011522621.2 | XP_011520923.1 | 6 - 7 | 6250 | 148 | |
XM_011522622.1 | XP_011520924.1 | 4 - 5 | 4954 | 148 | |
XM_011522623.2 | XP_011520925.1 | 7 - 8 | 3025 | 148 | |
XM_011522624.2 | XP_011520926.1 | 6 - 7 | 2363 | 148 | |
XM_011522625.2 | XP_011520927.1 | 5 - 6 | 2249 | 148 | |
XM_011522626.2 | XP_011520928.1 | 6 - 7 | 5814 | 148 | |
XM_011522627.2 | XP_011520929.1 | 6 - 7 | 5819 | 148 | |
XM_011522628.2 | XP_011520930.1 | 6 - 7 | 2883 | 148 | |
XM_011522629.2 | XP_011520931.1 | 6 - 7 | 2746 | 148 | |
XM_011522630.2 | XP_011520932.1 | 5 - 6 | 2084 | 148 | |
XM_011522631.2 | XP_011520933.1 | 4 - 5 | 1970 | 148 | |
XM_011522632.2 | XP_011520934.1 | 4 - 5 | 2401 | 148 | |
XM_017023596.1 | XP_016879085.1 | 7 - 8 | 3162 | 148 | |
GenBank mRNA | BC021854.1 | - | 1 - 2 | 90 | 148 |
U85647.1 | - | 5 - 6 | 2023 | 148 |