Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAPN15
Entrez Gene ID
6650
Gene Name
calpain 15
Gene Aliases
SOLH
Chromosome Location
Chr.16: 527795 - 554636 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005632.2 NP_005623.1 9 - 10 2702 107
XM_011522620.2 XP_011520922.1 10 - 11 6001 107
XM_011522621.2 XP_011520923.1 10 - 11 6929 107
XM_011522622.1 XP_011520924.1 8 - 9 5633 107
XM_011522623.2 XP_011520925.1 11 - 12 3704 107
XM_011522624.2 XP_011520926.1 10 - 11 3042 107
XM_011522625.2 XP_011520927.1 9 - 10 2928 107
XM_011522626.2 XP_011520928.1 10 - 11 6493 107
XM_011522627.2 XP_011520929.1 10 - 11 6498 107
XM_011522628.2 XP_011520930.1 10 - 11 3562 107
XM_011522629.2 XP_011520931.1 10 - 11 3425 107
XM_011522630.2 XP_011520932.1 9 - 10 2763 107
XM_011522631.2 XP_011520933.1 8 - 9 2649 107
XM_011522632.2 XP_011520934.1 8 - 9 3080 107
XM_017023596.1 XP_016879085.1 11 - 12 3841 107
GenBank mRNA BC021854.1 - 5 - 6 769 107
U85647.1 - 9 - 10 2702 107

View Full Product Details