The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177355.2 | NP_001170826.1 | 14 - 15 | 1481 | 127 |
NM_005823.5 | NP_005814.2 | 14 - 15 | 1541 | 127 | |
NM_013404.4 | NP_037536.2 | 13 - 14 | 1508 | 127 | |
XM_005255034.4 | XP_005255091.1 | 13 - 14 | 1787 | 127 | |
XM_011522348.2 | XP_011520650.1 | 14 - 15 | 1578 | 127 | |
XM_017022853.1 | XP_016878342.1 | 13 - 14 | 1725 | 127 | |
XM_017022854.1 | XP_016878343.1 | 13 - 14 | 1736 | 127 | |
XM_017022855.1 | XP_016878344.1 | 13 - 14 | 1634 | 127 | |
XM_017022856.1 | XP_016878345.1 | 13 - 14 | 1635 | 127 | |
XM_017022857.1 | XP_016878346.1 | 15 - 16 | 1655 | 127 | |
XM_017022858.1 | XP_016878347.1 | 14 - 15 | 1575 | 127 | |
GenBank mRNA | AF180951.1 | - | 3 - 4 | 485 | 127 |
AY743922.1 | - | 14 - 15 | 1541 | 127 | |
BC003512.1 | - | 13 - 14 | 1787 | 127 | |
BC009272.2 | - | 14 - 15 | 1480 | 127 | |
CB266931.1 | - | 3 - 4 | 429 | 127 | |
D49441.1 | - | 13 - 14 | 1465 | 127 | |
U40434.1 | - | 13 - 14 | 1490 | 127 |