The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003778.3 | NP_003769.1 | 2 - 3 | 277 | 80 |
XM_005247855.1 | XP_005247912.1 | 2 - 3 | 335 | 80 | |
XM_006713798.2 | XP_006713861.1 | 3 - 4 | 420 | 80 | |
XM_006713799.2 | XP_006713862.1 | 3 - 4 | 465 | 80 | |
XM_006713800.1 | XP_006713863.1 | 2 - 3 | 312 | 80 | |
XM_017007401.1 | XP_016862890.1 | 2 - 3 | 250 | 80 | |
GenBank mRNA | AF020920.1 | - | 1 - 2 | 168 | 80 |
AF038662.1 | - | 1 - 2 | 168 | 80 | |
AK314425.1 | - | 2 - 3 | 250 | 80 | |
BC062618.1 | - | 2 - 3 | 499 | 80 | |
BX647468.1 | - | 2 - 3 | 250 | 80 | |
CR749555.1 | - | 2 - 3 | 260 | 80 |