The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003778.3 | NP_003769.1 | 6 - 7 | 1212 | 121 |
NM_212543.1 | NP_997708.1 | 7 - 8 | 1295 | 121 | |
XM_005247855.1 | XP_005247912.1 | 6 - 7 | 1270 | 121 | |
XM_006713798.2 | XP_006713861.1 | 7 - 8 | 1355 | 121 | |
XM_006713799.2 | XP_006713862.1 | 7 - 8 | 1400 | 121 | |
XM_006713800.1 | XP_006713863.1 | 6 - 7 | 1247 | 121 | |
XM_006713801.2 | XP_006713864.1 | 8 - 9 | 1438 | 121 | |
XM_011513260.1 | XP_011511562.1 | 8 - 9 | 1367 | 121 | |
GenBank mRNA | AB024436.1 | - | 4 - 5 | 796 | 121 |
AF020920.1 | - | 5 - 6 | 1103 | 121 | |
AF022367.1 | - | 4 - 5 | 793 | 121 | |
AF038662.1 | - | 5 - 6 | 1103 | 121 | |
AK001006.1 | - | 7 - 8 | 1268 | 121 | |
AK314425.1 | - | 6 - 7 | 1185 | 121 | |
AY359008.1 | - | 4 - 5 | 911 | 121 | |
BC004523.2 | - | 5 - 6 | 961 | 121 | |
BC062618.1 | - | 6 - 7 | 1434 | 121 | |
BX647468.1 | - | 6 - 7 | 1185 | 121 | |
CR749555.1 | - | 6 - 7 | 1195 | 121 |