The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020754.3 | NP_065805.2 | 10 - 11 | 2172 | 82 |
XM_006713714.3 | XP_006713777.1 | 10 - 11 | 2196 | 82 | |
XM_017006955.1 | XP_016862444.1 | 9 - 10 | 2111 | 82 | |
GenBank mRNA | AB033030.1 | - | 10 - 11 | 2172 | 82 |
AK293726.1 | - | 10 - 11 | 1769 | 82 | |
BC112163.1 | - | 10 - 11 | 1667 | 82 | |
BC112165.1 | - | 10 - 11 | 1667 | 82 |