Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IZUMO4
Entrez Gene ID
113177
Gene Name
IZUMO family member 4
Gene Aliases
C19orf36, IMAGE:4215339
Chromosome Location
Chr.19: 2096848 - 2099590 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039846.1 NP_001034935.1 9 - 10 687 86
GenBank mRNA BI827974.1 - 9 - 10 542 86
BX378854.2 - 9 - 10 687 86
DQ196310.1 - 9 - 10 674 86
DQ424900.1 - 9 - 10 610 86

View Full Product Details