The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319.6 | NP_001310.3 | 1 - 2 | 255 | 95 |
XM_005259498.1 | XP_005259555.1 | 1 - 2 | 267 | 95 | |
XM_005259500.1 | XP_005259557.1 | 1 - 2 | 267 | 95 | |
GenBank mRNA | BC018693.2 | - | 1 - 2 | 228 | 95 |
BC018699.2 | - | 1 - 2 | 228 | 95 | |
CR994929.1 | - | 1 - 2 | 265 | 95 |