Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNMT3A
Entrez Gene ID
1788
Gene Name
DNA methyltransferase 3 alpha
Gene Aliases
DNMT3A2, M.HsaIIIA, TBRS
Chromosome Location
Chr.2: 25232961 - 25342590 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320893.1 NP_001307822.1 14 - 15 1905 65
NM_022552.4 NP_072046.2 19 - 20 2591 65
NM_153759.3 NP_715640.2 15 - 16 1875 65
NM_175629.2 NP_783328.1 19 - 20 2662 65
XM_005264175.4 XP_005264232.1 19 - 20 2525 65
XM_005264177.4 XP_005264234.1 14 - 15 1817 65
XM_011532662.2 XP_011530964.1 17 - 18 2527 65
XM_011532663.2 XP_011530965.1 20 - 21 2704 65
XM_011532666.2 XP_011530968.1 15 - 16 1890 65
XM_011532667.2 XP_011530969.1 14 - 15 1707 65
XM_017003526.1 XP_016859015.1 19 - 20 2633 65
XM_017003527.1 XP_016859016.1 14 - 15 1861 65
GenBank mRNA AF067972.2 - 19 - 20 2553 65
AF331856.1 - 19 - 20 2541 65
AF480163.1 - 15 - 16 1871 65
AK025230.1 - 9 - 10 915 65
BC018214.1 - 4 - 5 444 65
BC023612.2 - 5 - 6 643 65
BC043617.1 - 19 - 20 2561 65

View Full Product Details