The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320893.1 | NP_001307822.1 | 3 - 4 | 590 | 65 |
NM_022552.4 | NP_072046.2 | 8 - 9 | 1276 | 65 | |
NM_153759.3 | NP_715640.2 | 4 - 5 | 560 | 65 | |
NM_175629.2 | NP_783328.1 | 8 - 9 | 1347 | 65 | |
NR_135490.1 | 8 - 9 | 1347 | 65 | ||
XM_005264175.4 | XP_005264232.1 | 8 - 9 | 1210 | 65 | |
XM_005264177.4 | XP_005264234.1 | 3 - 4 | 502 | 65 | |
XM_011532662.2 | XP_011530964.1 | 6 - 7 | 1212 | 65 | |
XM_011532663.2 | XP_011530965.1 | 9 - 10 | 1389 | 65 | |
XM_011532664.2 | XP_011530966.1 | 8 - 9 | 1287 | 65 | |
XM_011532666.2 | XP_011530968.1 | 4 - 5 | 575 | 65 | |
XM_011532667.2 | XP_011530969.1 | 3 - 4 | 392 | 65 | |
XM_017003526.1 | XP_016859015.1 | 8 - 9 | 1318 | 65 | |
XM_017003527.1 | XP_016859016.1 | 3 - 4 | 546 | 65 | |
GenBank mRNA | AB208833.1 | - | 8 - 9 | 1246 | 65 |
AF067972.2 | - | 8 - 9 | 1238 | 65 | |
AF331856.1 | - | 8 - 9 | 1226 | 65 | |
AF480163.1 | - | 4 - 5 | 556 | 65 | |
BC043617.1 | - | 8 - 9 | 1246 | 65 | |
BM470515.1 | - | - | 572 | 65 |