Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NR1H2
Entrez Gene ID
7376
Gene Name
nuclear receptor subfamily 1 group H member 2
Gene Aliases
LXR-b, LXRB, NER, NER-I, RIP15, UNR
Chromosome Location
Chr.19: 50376423 - 50383028 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256647.1 NP_001243576.1 1 - 2 149 71
NM_007121.5 NP_009052.3 1 - 2 149 71
GenBank mRNA AK091535.1 - 5 - 6 747 71
AK290855.1 - 1 - 2 80 71
AK297978.1 - 1 - 2 114 71
AL598714.1 - 1 - 2 127 71
BC033500.1 - 1 - 2 67 71
BC047750.2 - 1 - 2 103 71
BC074500.1 - 1 - 2 84 71
CR749648.1 - 1 - 2 70
DA053472.1 - 1 - 2 149 71
U07132.1 - 1 - 2 92 71

View Full Product Details